4-Benzylpyrrolidin-2-one

Supplier: Matrix Scientific

063314-500MG
101925-530EA 183.08 USD
101925-530
4-Benzylpyrrolidin-2-one
4-Benzylpyrrolidin-2-one

Hazard: Irritant

Formula: C₁₁H₁₃NO
MDL Number: MFCD09864502
CAS Number: 30200-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR