2-Azaspiro[4.6]undecan-3-one

Supplier: Matrix Scientific

063313-500MG
101925-528EA 364.62 USD
101925-528
2-Azaspiro[4.6]undecan-3-one
2-Azaspiro[4.6]undecan-3-one

Hazard: Irritant

Formula: C₁₀H₁₇NO
MDL Number: MFCD09864499
CAS Number: 134617-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR