4-Propylpyrrolidin-2-one

Supplier: Matrix Scientific

063309-500MG
101925-520EA 484.62 USD
101925-520
4-Propylpyrrolidin-2-one
4-Propylpyrrolidin-2-one

Hazard: Irritant

Formula: C₇H₁₃NO
MDL Number: MFCD15146418
CAS Number: 89895-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR