tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate

063306-500MG
101925-514EA 364.62 USD
101925-514
tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate
tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₂N₂O₃
MW: 254.33 g/mol
MDL Number: MFCD11111226
CAS Number: 169206-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR