4-Benzyl-1,4-oxazepan-3-one

Supplier: Matrix Scientific

063302-500MG
101925-510EA 183.08 USD
101925-510
4-Benzyl-1,4-oxazepan-3-one
4-Benzyl-1,4-oxazepan-3-one

Hazard: Irritant

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD15146415
CAS Number: 109069-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR