Ethyl-5-propyl-1H-pyrazole-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 5-Propylpyrazole-3-carboxylate

063298-500MG
101925-502EA 484.62 USD
101925-502
Ethyl-5-propyl-1H-pyrazole-3-carboxylate
Ethyl-5-propyl-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
MDL Number: MFCD08700607
CAS Number: 92945-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR