6-Azaspiro[3.5]nonan-5-one

Supplier: Matrix Scientific

063294-500MG
101925-492EA 484.62 USD
101925-492
6-Azaspiro[3.5]nonan-5-one
6-Azaspiro[3.5]nonan-5-one

Hazard: Irritant

Formula: C₈H₁₃NO
MDL Number: MFCD15146412
CAS Number: 1217862-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR