4-Azaspiro[2.4]heptan-5-one

Supplier: Matrix Scientific

063293-500MG
101925-490EA 461.54 USD
101925-490
4-Azaspiro[2.4]heptan-5-one
4-Azaspiro[2.4]heptan-5-one

Hazard: Irritant

Formula: C₆H₉NO
MDL Number: MFCD15146411
CAS Number: 308266-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR