5-Chloro-3-methyl-1H-indole-2-carbaldehyde

Supplier: Matrix Scientific

063281-500MG
101925-466EA 183.08 USD
101925-466
5-Chloro-3-methyl-1H-indole-2-carbaldehyde
5-Chloro-3-methyl-1H-indole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₈ClNO
MDL Number: MFCD08059791
CAS Number: 40731-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR