3-(1H-Pyrazol-3-yl)pyridine

Supplier: Matrix Scientific

063280-500MG
101925-464EA 484.62 USD
101925-464
3-(1H-Pyrazol-3-yl)pyridine
3-(1H-Pyrazol-3-yl)pyridine

Hazard: Irritant

Formula: C₈H₇N₃
MW: 145.16 g/mol
MDL Number: MFCD03407951
CAS Number: 45887-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR