3-(2-Furyl)-1H-pyrazole

Supplier: Matrix Scientific

063279-500MG
101925-462EA 243.08 USD
101925-462
3-(2-Furyl)-1H-pyrazole
3-(2-Furyl)-1H-pyrazole

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD00973923
CAS Number: 32332-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR