2-Bromo-1-cyclopropylethanone ≥90%

Supplier: Matrix Scientific

063275-25G
101925-456EA 421.54 USD
101925-456
2-Bromo-1-cyclopropylethanone ≥90%
2-Bromo-1-cyclopropylethanone

Hazard: Irritant/Lachrymator

Formula: C₅H₇BrO
Boiling Pt: 191 °C
Density: 1.669
Flash Pt: 92 °C
MDL Number: MFCD08460238
CAS Number: 69267-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR