6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine

Supplier: Matrix Scientific

Synonyms: 6-Chloro-3-methyl-[1,2,4]triazolo[4,3-b]pyridazine

063263-500MG
101925-430EA 387.69 USD
101925-430
6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine
6-Chloro-3-methyl[1,2,4]triazolo[4,3-b]pyridazine

Hazard: Irritant

Formula: C₆H₅ClN₄
MW: 168.59 g/mol
Melting Pt: 104…105 °C
MDL Number: MFCD00975127
CAS Number: 7197-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR