[1-(Pyridin-3-ylmethyl)piperidin-3-yl]methanol

Supplier: Matrix Scientific

063259-500MG
101925-422EA 387.69 USD
101925-422
[1-(Pyridin-3-ylmethyl)piperidin-3-yl]methanol
[1-(Pyridin-3-ylmethyl)piperidin-3-yl]methanol

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD00978389
CAS Number: 331978-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR