2-Azido-N-methylacetamide

Supplier: Matrix Scientific

063255-500MG
101925-416EA 387.69 USD
101925-416
2-Azido-N-methylacetamide
2-Azido-N-methylacetamide

Hazard: Irritant

Formula: C₃H₆N₄O
MDL Number: MFCD13248783
CAS Number: 98025-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR