(2-Furylmethyl)(1H-indol-6-ylmethyl)amine

Supplier: Matrix Scientific

063252-500MG
101925-410EA 484.62 USD
101925-410
(2-Furylmethyl)(1H-indol-6-ylmethyl)amine
(2-Furylmethyl)(1H-indol-6-ylmethyl)amine

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD13248782
CAS Number: 1030430-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR