N-(1H-Indol-6-ylmethyl)cyclopropanamine

Supplier: Matrix Scientific

063251-500MG
101925-408EA 444.62 USD
101925-408
N-(1H-Indol-6-ylmethyl)cyclopropanamine
N-(1H-Indol-6-ylmethyl)cyclopropanamine

Hazard: Irritant

Formula: C₁₂H₁₄N₂
MDL Number: MFCD11556359
CAS Number: 1079-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR