(2-Furylmethyl)(1H-indol-5-ylmethyl)amine

Supplier: Matrix Scientific

063250-500MG
101925-406EA 484.62 USD
101925-406
(2-Furylmethyl)(1H-indol-5-ylmethyl)amine
(2-Furylmethyl)(1H-indol-5-ylmethyl)amine

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD13248780
CAS Number: 946700-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR