2-Chloro-5-[(methylthio)methyl]pyridine

Supplier: Matrix Scientific

063246-500MG
101925-400EA 484.62 USD
101925-400
2-Chloro-5-[(methylthio)methyl]pyridine
2-Chloro-5-[(methylthio)methyl]pyridine

Hazard: Irritant

Formula: C₇H₈ClNS
MDL Number: MFCD12823579
CAS Number: 1021870-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR