2-Chloro-5-(1H-imidazol-1-ylmethyl)pyridine

Supplier: Matrix Scientific

063245-500MG
101925-398EA 364.62 USD
101925-398
2-Chloro-5-(1H-imidazol-1-ylmethyl)pyridine
2-Chloro-5-(1H-imidazol-1-ylmethyl)pyridine

Hazard: Irritant

Formula: C₉H₈ClN₃
MW: 193.64 g/mol
MDL Number: MFCD04124604
CAS Number: 230617-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR