1-Isopropyl-1H-indole-6-carbaldehyde

Supplier: Matrix Scientific

063244-500MG
101925-396EA 290.77 USD
101925-396
1-Isopropyl-1H-indole-6-carbaldehyde
1-Isopropyl-1H-indole-6-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₃NO
MDL Number: MFCD11877854
CAS Number: 921602-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR