2-Chloro-N-[2-(3-methoxyphenyl)ethyl]acetamide

Supplier: Matrix Scientific

063241-500MG
101925-392EA 364.62 USD
101925-392
2-Chloro-N-[2-(3-methoxyphenyl)ethyl]acetamide
2-Chloro-N-[2-(3-methoxyphenyl)ethyl]acetamide

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₂
MDL Number: MFCD01733621
CAS Number: 34162-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR