1-(Chloroacetyl)-4-(4-chlorophenyl)piperazine
Supplier: Matrix Scientific
063239-500MG
101925-388EA
243.08
USD
101925-388
1-(Chloroacetyl)-4-(4-chlorophenyl)piperazine
1-(Chloroacetyl)-4-(4-chlorophenyl)piperazine
Hazard: Irritant
Formula:
C₁₂H₁₄Cl₂N₂O |
MDL Number:
MFCD01257158 CAS Number: 60121-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...