2-Chloro-N-(2-methyl-5-nitrophenyl)acetamide

Supplier: Matrix Scientific

063235-500MG
101925-380EA 243.08 USD
101925-380
2-Chloro-N-(2-methyl-5-nitrophenyl)acetamide
2-Chloro-N-(2-methyl-5-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₉H₉ClN₂O₃
MDL Number: MFCD00031298
CAS Number: 35588-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR