2-Chloro-N-(4-chloro-2-methylphenyl)acetamide

Supplier: Matrix Scientific

Synonyms: N1-(4-Chloro-2-methylphenyl)-2-chloroacetamide

063231-500MG
101925-372EA 243.08 USD
101925-372
2-Chloro-N-(4-chloro-2-methylphenyl)acetamide
2-Chloro-N-(4-chloro-2-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₉Cl₂NO
MW: 218.08 g/mol
MDL Number: MFCD00018906
CAS Number: 62593-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR