2-Chloro-N-propylacetamide

Supplier: Matrix Scientific

063228-500MG
101925-366EA 243.08 USD
101925-366
2-Chloro-N-propylacetamide
2-Chloro-N-propylacetamide

Hazard: Irritant

Formula: C₅H₁₀ClNO
MDL Number: MFCD01355490
CAS Number: 13916-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR