2-(Isothiocyanatomethyl)furan

Supplier: Matrix Scientific

063223-500MG
101925-356EA 146.15 USD
101925-356
2-(Isothiocyanatomethyl)furan
2-(Isothiocyanatomethyl)furan

Hazard: Irritant

Formula: C₆H₅NOS
MDL Number: MFCD00014112
CAS Number: 4650-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR