Methyl (4-isothiocyanatophenyl)acetate

Supplier: Matrix Scientific

063218-500MG
101925-348EA 364.62 USD
101925-348
Methyl (4-isothiocyanatophenyl)acetate
Methyl (4-isothiocyanatophenyl)acetate

Hazard: Irritant

Formula: C₁₀H₉NO₂S
MDL Number: MFCD03759881
CAS Number: 77055-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR