4-Chloro-2,5-dimethoxyphenyl isothiocyanate

Supplier: Matrix Scientific

063217-500MG
101925-346EA 146.15 USD
101925-346
4-Chloro-2,5-dimethoxyphenyl isothiocyanate
4-Chloro-2,5-dimethoxyphenyl isothiocyanate

Hazard: Irritant

Formula: C₉H₈ClNO₂S
MDL Number: MFCD00046809
CAS Number: 306935-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR