(4-Isothiocyanatophenyl)phenylamine

Supplier: Matrix Scientific

063216-500MG
101925-344EA 364.62 USD
101925-344
(4-Isothiocyanatophenyl)phenylamine
(4-Isothiocyanatophenyl)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₀N₂S
MDL Number: MFCD03759880
CAS Number: 23246-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR