1-(3-Fluoro-4-pyrrolidin-1-ylphenyl)ethanone

Supplier: Matrix Scientific

063208-500MG
101925-338EA 364.62 USD
101925-338
1-(3-Fluoro-4-pyrrolidin-1-ylphenyl)ethanone
1-(3-Fluoro-4-pyrrolidin-1-ylphenyl)ethanone

Hazard: Irritant

Formula: C₁₂H₁₄FNO
MDL Number: MFCD02633467
CAS Number: 385380-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR