1-[3-Fluoro-4-(3-methylpiperidin-1-yl)phenyl]ethanone

Supplier: Matrix Scientific

063206-500MG
101925-334EA 364.62 USD
101925-334
1-[3-Fluoro-4-(3-methylpiperidin-1-yl)phenyl]ethanone
1-[3-Fluoro-4-(3-methylpiperidin-1-yl)phenyl]ethanone

Hazard: Irritant

Formula: C₁₄H₁₈FNO
MDL Number: MFCD11136772
CAS Number: 1019547-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR