Methyl [1-(4-methoxybenzoyl)piperidin-4-yl]acetate

Supplier: Matrix Scientific

063197-500MG
101925-318EA 484.62 USD
101925-318
Methyl [1-(4-methoxybenzoyl)piperidin-4-yl]acetate
Methyl [1-(4-methoxybenzoyl)piperidin-4-yl]acetate

Hazard: Irritant

Formula: C₁₆H₂₁NO₄
MDL Number: MFCD13248768
CAS Number: 952934-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR