2-(tert-Butylamino)-5-nitrobenzonitrile

Supplier: Matrix Scientific

063192-500MG
101925-308EA 364.62 USD
101925-308
2-(tert-Butylamino)-5-nitrobenzonitrile
2-(tert-Butylamino)-5-nitrobenzonitrile

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₂
MDL Number: MFCD09455382
CAS Number: 945299-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR