Methyl-2-anilino-4-oxo-4,5-dihydrothiophene-3-carboxylate

Supplier: Matrix Scientific

063188-500MG
101925-304EA 290.77 USD
101925-304
Methyl-2-anilino-4-oxo-4,5-dihydrothiophene-3-carboxylate
Methyl-2-anilino-4-oxo-4,5-dihydrothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃S
MDL Number: MFCD02221506
CAS Number: 425650-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR