2-Methyl-3,4-dihydro-2H-thieno[2,3-e][1,2]thiazin-4-ol-1,1-dioxide

Supplier: Matrix Scientific

063184-500MG
101925-300EA 484.62 USD
101925-300
2-Methyl-3,4-dihydro-2H-thieno[2,3-e][1,2]thiazin-4-ol-1,1-dioxide
2-Methyl-3,4-dihydro-2H-thieno[2,3-e][1,2]thiazin-4-ol-1,1-dioxide

Hazard: Irritant

Formula: C₇H₉NO₃S₂
MDL Number: MFCD13248763
CAS Number: 1030431-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR