3-Amino-2-thioxo-2,3-dihydroquinazolin-4(1H)-one

Supplier: Matrix Scientific

063182-500MG
101925-298EA 290.77 USD
101925-298
3-Amino-2-thioxo-2,3-dihydroquinazolin-4(1H)-one
3-Amino-2-thioxo-2,3-dihydroquinazolin-4(1H)-one

Hazard: Irritant

Formula: C₈H₇N₃OS
MDL Number: MFCD03283476
CAS Number: 16951-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR