N'-(Chloroacetyl)-2,2-diphenylacetohydrazide

Supplier: Matrix Scientific

063177-500MG
101925-290EA 243.08 USD
101925-290
N'-(Chloroacetyl)-2,2-diphenylacetohydrazide
N'-(Chloroacetyl)-2,2-diphenylacetohydrazide

Hazard: Irritant

Formula: C₁₆H₁₅ClN₂O₂
MDL Number: MFCD08547719
CAS Number: 909370-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR