2-(Diaminomethylene)-5,5-dimethylcyclohexane-1,3-dione

Supplier: Matrix Scientific

063171-500MG
101925-280EA 364.62 USD
101925-280
2-(Diaminomethylene)-5,5-dimethylcyclohexane-1,3-dione
2-(Diaminomethylene)-5,5-dimethylcyclohexane-1,3-dione

Hazard: Irritant

Formula: C₉H₁₄N₂O₂
MDL Number: MFCD00461409
CAS Number: 384811-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR