[2-(4H-1,2,4-Triazol-4-yl)phenoxy]acetic acid

Supplier: Matrix Scientific

063160-500MG
101925-260EA 364.62 USD
101925-260
[2-(4H-1,2,4-Triazol-4-yl)phenoxy]acetic acid
[2-(4H-1,2,4-Triazol-4-yl)phenoxy]acetic acid

Hazard: Irritant

Formula: C₁₀H₉N₃O₃
MDL Number: MFCD13248752
CAS Number: 1092250-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR