[3-(5-Isopropyl-1,3-benzoxazol-2-yl)-2-methylphenyl]amine

Supplier: Matrix Scientific

063159-500MG
101925-258EA 364.62 USD
101925-258
[3-(5-Isopropyl-1,3-benzoxazol-2-yl)-2-methylphenyl]amine
[3-(5-Isopropyl-1,3-benzoxazol-2-yl)-2-methylphenyl]amine

Hazard: Irritant

Formula: C₁₇H₁₈N₂O
MDL Number: MFCD03032211
CAS Number: 443292-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR