4-Bromo-2-(4-fluorophenyl)-6-methyl-2H-1,2,3-benzotriazol-5-amine

Supplier: Matrix Scientific

063158-500MG
101925-256EA 243.08 USD
101925-256
4-Bromo-2-(4-fluorophenyl)-6-methyl-2H-1,2,3-benzotriazol-5-amine
4-Bromo-2-(4-fluorophenyl)-6-methyl-2H-1,2,3-benzotriazol-5-amine

Hazard: Irritant

Formula: C₁₃H₁₀BrFN₄
MDL Number: MFCD13187627
CAS Number: 952948-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR