5-(2-Bromophenyl)-2-furonitrile

Supplier: Matrix Scientific

063154-500MG
101925-248EA 364.62 USD
101925-248
5-(2-Bromophenyl)-2-furonitrile
5-(2-Bromophenyl)-2-furonitrile

Hazard: Irritant

Formula: C₁₁H₆BrNO
MDL Number: MFCD07360126
CAS Number: 88649-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR