2-(2,4-Dichloro-6-methylphenoxy)acetamide

Supplier: Matrix Scientific

063148-500MG
101925-236EA 183.08 USD
101925-236
2-(2,4-Dichloro-6-methylphenoxy)acetamide
2-(2,4-Dichloro-6-methylphenoxy)acetamide

Hazard: Irritant

Formula: C₉H₉Cl₂NO₂
MDL Number: MFCD13187467
CAS Number: 501008-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR