2-(4-tert-Butylphenoxy)acetamide

Supplier: Matrix Scientific

063147-500MG
101925-234EA 183.08 USD
101925-234
2-(4-tert-Butylphenoxy)acetamide
2-(4-tert-Butylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD01610040
CAS Number: 28329-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR