2,3-Dibromo-5-ethoxy-6-hydroxybenzaldehyde oxime

Supplier: Matrix Scientific

063146-500MG
101925-232EA 146.15 USD
101925-232
2,3-Dibromo-5-ethoxy-6-hydroxybenzaldehyde oxime
2,3-Dibromo-5-ethoxy-6-hydroxybenzaldehyde oxime

Hazard: Irritant

Formula: C₉H₉Br₂NO₃
MDL Number: MFCD11519737
CAS Number: 1030445-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR