5-Chloro-1-naphthoic acid

Supplier: Matrix Scientific

063144-500MG
101925-228EA 335.38 USD
101925-228
5-Chloro-1-naphthoic acid
5-Chloro-1-naphthoic acid

Hazard: Irritant

Formula: C₁₁H₇ClO₂
CAS Number: 16650-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR