(2Z,3Z)-2-(Hydroxyimino)-N-phenyl-3-(phenylhydrazono)butanamide

Supplier: Matrix Scientific

063139-500MG
101925-218EA 183.08 USD
101925-218
(2Z,3Z)-2-(Hydroxyimino)-N-phenyl-3-(phenylhydrazono)butanamide
(2Z,3Z)-2-(Hydroxyimino)-N-phenyl-3-(phenylhydrazono)butanamide

Hazard: Irritant

Formula: C₁₆H₁₆N₄O₂
MDL Number: MFCD13248751
CAS Number: 6030-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR