3-(5-Amino-2H-1,2,3-benzotriazol-2-yl)benzoic acid

Supplier: Matrix Scientific

063137-500MG
101925-214EA 484.62 USD
101925-214
3-(5-Amino-2H-1,2,3-benzotriazol-2-yl)benzoic acid
3-(5-Amino-2H-1,2,3-benzotriazol-2-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₀N₄O₂
MDL Number: MFCD11556629
CAS Number: 873994-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR