5-Bromo-2-methoxy-3-methylbenzamide

Supplier: Matrix Scientific

063135-500MG
101925-210EA 183.08 USD
101925-210
5-Bromo-2-methoxy-3-methylbenzamide
5-Bromo-2-methoxy-3-methylbenzamide

Hazard: Irritant

Formula: C₉H₁₀BrNO₂
MDL Number: MFCD01667007
CAS Number: 58708-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR